missing translation for 'onlineSavingsMsg'
Learn More

OICR-0547, MedChemExpress

Product Code. 16457216
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457216 100 mg 100mg
16467248 10 mg 10mg
16439777 5 mg 5mg
16433566 25 mg 25mg
16433267 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16457216 Supplier Medchem Express Supplier No. HY16994

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OICR-0547 is a closely related derivative of OICR-9429. OICR-9429 is a novel small-molecule antagonist of the Wdr5-MLL interaction, while OICR-0547 is an inactive control compound that no longer binds to WDR5.

OICR-0547 is a closely related derivative of OICR-9429. OICR-9429 is a novel small-molecule antagonist of the Wdr5-MLL interaction, while OICR-0547 is an inactive control compound that no longer binds to WDR5.

Specifications

Chemical Name or Material OICR-0547
Physical Form Solid
CAS 1801873-49-3
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C28H29F3N4O4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (184.31 mM)
SMILES O=C(C1=CNC(C=C1C(F)(F)F)=O)NC2=CC(C3=CC(CN4CCOCC4)=CC=C3)=CC=C2N5CCOCC5
Molecular Weight (g/mol) 542.55
Formula Weight 542.55
Percent Purity 99.93%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.