missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OICR-0547, MedChemExpress
Description
OICR-0547 is a closely related derivative of OICR-9429. OICR-9429 is a novel small-molecule antagonist of the Wdr5-MLL interaction, while OICR-0547 is an inactive control compound that no longer binds to WDR5.
Specifications
Specifications
| Chemical Name or Material | OICR-0547 |
| Physical Form | Solid |
| CAS | 1801873-49-3 |
| Color | Light Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C28H29F3N4O4 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : ≥ 100 mg/mL (184.31 mM) |
| SMILES | O=C(C1=CNC(C=C1C(F)(F)F)=O)NC2=CC(C3=CC(CN4CCOCC4)=CC=C3)=CC=C2N5CCOCC5 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?