Learn More
Welch™ Oil Mist Separator for CRVpro Direct Drive Rotary Vane Vacuum Pump
For use with CRVpro Direct Drive Rotary Vane Vacuum Pump
Brand:  Welch™ 320015
Additional Details : Weight : 0.99000kg
Product Code. 10211865
  • € 294.68 / Each
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
Description
Description
  • Outlet oil separator removes oil particles from exhaust stream, minimizing oil loss to atmosphere
Specifications
Specifications
Outlet Oil Mist Separator
Removes oil particles from exhaust stream
Rotary vane pump