Learn More
Kern Sohn OIML Class E1 Weight Set, Wooden Box
Click to view available options
Weight (Metric):
1 g to 1 kg
1 g to 10 kg
1 g to 2 kg
1 g to 5 kg
1 mg to 1 kg
1 mg to 10 kg
1 mg to 100 g
1 mg to 2 kg
1 mg to 200 g
1 mg to 5 kg
1 mg to 50 g
1 mg to 500 g
1 to 100 g
1 to 200 g
1 to 50 g
1 to 500 g
1 to 500 mg
Unit Size:
1 set
Description
- Set of weights
- Polished stainless steel
- Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic case
Specifications
Specifications
| Classification | E1 |
| Material | Stainless Steel |
| Case Material | Wood |
| Weight (Metric) | 1 to 100 g |