Learn More
Kern Sohn OIML Class E1 Wire Test Weight
Product Code. 17361863
Encompass_Preferred
Click to view available options
Weight (Metric):
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
Each
Product Code. 17361863
Brand: Kern Sohn 30831
Only null left
Add to basket
Add to basket
Single test weight constructed of stainless steel.
  • Wire shape
  • Stainless steel
Specifications
Classification E1
Material Stainless Steel
Weight (Metric) 1 mg
Tolerance ± 0.003 mg