Learn More
Kern Sohn OIML Class F2 Knob Test Weight
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
20 kg
50 kg
Unit Size:
Each
Description
- Knob shape
- Finely turned stainless steel
Specifications
Specifications
| Classification | F2 |
| Material | Stainless Steel |
| Weight (Metric) | 50 kg |