Learn More
Kern Sohn OIML Class M1 Flat Sheet Test Weight
Product Code. 17342453
Encompass_Preferred
Click to view available options
Weight (Metric):
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
Each
Product Code. 17342453
Brand: Kern Sohn T34805A
Only null left
Add to basket
Add to basket
Platelet test weight constructed of stainless steel.
  • Flat polygonal sheet
  • Polished stainless steel
Specifications
Classification M1
Material Stainless Steel
Weight (Metric) 20 mg
Tolerance ± 0.3 mg