Learn More
Kern Sohn OIML Class M1 Slot Test Weight
Product Code. 17382323
Encompass_Preferred
Click to view available options
Weight (Metric):
1 cN
1 g
10 cN
10 g
100 N
100 g
2 N
2 cN
2 g
2 kg
20 g
200 g
5 N
5 cN
5 g
5 kg
50 N
50 cN
50 g
500 g
Unit Size:
Each
Product Code. 17382323
Brand: Kern Sohn 347495
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Single test weight constructed of brass.
  • Slot shape
  • Finely turned brass
Specifications
Classification M1
Material Brass
Weight (Metric) 500 g