Learn More
Kern Sohn OIML Class Weight Set, Platelet
Product Code. 17372183
Encompass_Preferred
Click to view available options
Classification:
E2
F1
F2
M1
Unit Size:
1 set
Each
Product Code. 17372183
Brand: Kern Sohn 33822
Only null left
Add to basket
Add to basket
Weight set constructed of polished stainless steel.
  • Polished stainless steel
Specifications
Classification F2
Material Stainless Steel
Case Material Plastic
Weight (Metric) 1 to 500 mg