missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OIP106 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98550DL594
This item is not returnable.
View return policy
Description
OIP106 Polyclonal antibody specifically detects OIP106 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| OIP106 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 22906 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| 106 kDa O-GlcNAc transferase-interacting protein, KIAA1042OGT(O-Glc-NAc transferase)-interacting protein 106 KDa, OIP106OGT(O Glc NAc transferase) interacting protein 106 KDa, trafficking kinesin-binding protein 1, trafficking protein, kinesin binding 1 | |
| Produced in rabbits immunized with E. coli-derived Human OIP106 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction