missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OLAH Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98493AF532
This item is not returnable.
View return policy
Description
OLAH Polyclonal antibody specifically detects OLAH in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| OLAH | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55301 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| EC 3.1.2.14, FLJ11106, medium chain, MGC51852, oleoyl-ACP hydrolaseAURA1, SAST, THEDC1, thioesterase domain containing 1, Thioesterase II | |
| Produced in rabbits immunized with E. coli-derived Human OLAH fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction