Learn More
Oleandrin, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
Description
Oleandrin (PBI-05204) inhibits the Na+, K+-ATPase activity with an IC50 of 620 nM.
Specifications
Specifications
| Chemical Name or Material | Oleandrin |
| CAS | 465-16-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C32H48O9 |
| Quantity | 25 mg |
| Synonym | PBI-05204 |
| Solubility Information | DMSO : 100 mg/mL (173.39 mM; Need ultrasonic) |
| SMILES | O=C1OCC([C@H]2[C@@H](OC(C)=O)C[C@]3(O)[C@]4([H])CC[C@]5([H])C[C@@H](O[C@H]6C[C@@H]([C@H]([C@H](C)O6)O)OC)CC[C@]5(C)[C@@]4([H])CC[C@]23C)=C1 |
| Molecular Weight (g/mol) | 576.72 |
| Show More |
Research purposes only