missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OLFM4 Antibody (12), Alexa Fluor™ 750 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06415AF750
This item is not returnable.
View return policy
Description
OLFM4 Monoclonal antibody specifically detects OLFM4 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
| OLFM4 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Antiapoptotic protein GW112, bA209J19.1, GC1, G-CSF-stimulated clone 1 protein, GW112hOLfD, hGC-1, KIAA4294, olfactomedin 4, olfactomedin-4, OlfD, OLM4, UNQ362 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human OLFM4 (NP_006409.3; Met 1-Gln 510). | |
| 0.1 mL | |
| Cancer | |
| 10562 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, ELISA, Immunofluorescence | |
| 12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction