missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olig2 Antibody (OLIG2/7366R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 287.00 - € 519.00
Specifications
| Antigen | Olig2 |
|---|---|
| Clone | OLIG2/7366R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663415
|
Novus Biologicals
NBP3-20429-20ug |
20 μg |
€ 287.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18652405
|
Novus Biologicals
NBP3-20429-100ug |
100 μg |
€ 519.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Olig2 Monoclonal antibody specifically detects Olig2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Olig2 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Stem Cell Markers, Transcription Factors and Regulators | |
| 10mM PBS with 0.05% BSA | |
| 10215 | |
| IgG κ | |
| Protein A or G purified |
| OLIG2/7366R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BHLHB1, BHLHE19, Class B basic helix-loop-helix protein 1, Class E basic helix-loop-helix protein 19, human protein kinase C-binding protein RACK17, OLIGO2, oligodendrocyte lineage transcription factor 2, oligodendrocyte transcription factor 2, oligodendrocyte-specific bHLH transcription factor 2, protein kinase C binding protein 2, Protein kinase C-binding protein 2, Protein kinase C-binding protein RACK17, RACK17helix-loop-helix protein, class B, 1 | |
| Recombinant fragment (around aa200-300) of human Olig2 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title