missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OLIG2 Recombinant Rabbit Monoclonal Antibody (1P8G4)

OLIG2 Recombinant Rabbit Monoclonal Antibody (ARC1032)
Brand: Invitrogen™ MA535533
This item is not returnable.
View return policy
Description
OLIG2 Recombinant Monoclonal Antibody for Western Blot
This gene encodes a basic helix-loop-helix transcription factor which is expressed in oligodendroglial tumors of the brain. The protein is an essential regulator of ventral neuroectodermal progenitor cell fate. The gene is involved in a chromosomal translocation t(14;21)(q11.2;q22) associated with T-cell acute lymphoblastic leukemia. Its chromosomal location is within a region of chromosome 21 which has been suggested to play a role in learning deficits associated with Down syndrome. Tissue specificity: Expressed in the brain, in oligodendrocytes. Strongly expressed in oligodendrogliomas, while expression is weak to moderate in astrocytomas. Expression in glioblastomas highly variable.
Specifications
| OLIG2 | |
| Recombinant Monoclonal | |
| 0.43 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9EQW6 | |
| OLIG2 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human Olig2 (Q13516). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1P8G4 | |
| Unconjugated | |
| OLIG2 | |
| AI604895; basic domain, helix-loop-helix protein, class B, 1; Basic helix loop helix protein class B1 (bHLHB1); basic helix-loop-helix protein 19 (bHLHe19); bHLH transcription factor; BHLHB1; bHLHe19; Class B basic helix-loop-helix protein 1; class E basic helix-loop-helix protein 19; human protein kinase C-binding protein RACK17; Olg-2; OLIG2; olig-2; Oligo2; Oligo2 antibody; oligodendrocyte lineage transcription factor 2; Oligodendrocyte specific bHLH transcription factor 2; oligodendrocyte transcription factor 2; oligodendrocyte-specific bHLH transcription factor 2; PRKCBP2; Protein kinase C-binding protein 2; Protein kinase C-binding protein 2 (PRKCBP2); Protein kinase C-binding protein RACK17; RACK17; RK17; wu:fc64g09 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 304103, 50913 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction