missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OLIG3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110060
This item is not returnable.
View return policy
Description
Immunogen sequence: TVGHSAGHPA HAANSVHPVH PILGGALSSG NASSPLSAAS LPAIGTIRPP HSLLKAPSTP PALQLGSGFQ HWAGLPCPCT ICQMPPPPHL SALSTANMAR LSAESKDLLK.
Oligodendrocyte transcription factor 3 (OLIG3) is a gene located on chromosome 6q23.2, encoding a basic helix-loop-helix (bHLH) transcription factor that is involved in the regulation of neural development. OLIG3 is a member of the OLIG family, which plays a significant role in the differentiation and maturation of oligodendrocytes, the myelinating cells of the central nervous system. This transcription factor is crucial for the specification and development of certain neuronal subtypes, contributing to the formation and maintenance of the central nervous system's architecture. OLIG3's activity influences gene expression related to neural proliferation, survival, and differentiation. Although less studied than OLIG1 and OLIG2, OLIG3's expression patterns suggest its involvement in neural patterning and potential roles in neuronal disorders. Research continues to explore the mechanisms through which OLIG3 affects neural development and its implications in neuropathologies and regenerative medicine.
Specifications
| OLIG3 | |
| Polyclonal | |
| Unconjugated | |
| OLIG3 | |
| basic helix-loop-helix domain-containing class B protein 7; Bhlhb7; bHLHe20; bHLHe20oligo3; Class B basic helix-loop-helix protein 7; class E basic helix-loop-helix protein 20; OLIG3; Oligo3; oligodendrocyte transcription factor 2; oligodendrocyte transcription factor 2; oligodendrocyte transcription factor 3; oligodendrocyte transcription factor 3; Oligodendrocyte-specific bHLH transcription factor 3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 293012, 94222 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.43 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6PFG8 | |
| OLIG3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 163-272 of human OLIG3 (NP_786923.1). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction