missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oligodendrocyte Marker O1 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1327V-100UG
This item is not returnable.
View return policy
Description
Oligodendrocyte Marker O1 Monoclonal specifically detects Oligodendrocyte Marker O1 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry.
Specifications
| Oligodendrocyte Marker O1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| IgM |
| Flow Cytometry | |
| O1 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Bovine brain corpus callosum white matter | |
| RUO | |
| Detects human, mouse, rat and chicken Oligodendrocyte Marker O1. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction