missing translation for 'onlineSavingsMsg'
Learn More

Olmutinib, MedChemExpress

Product Code. 16477868
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16477868

Brand: Medchem Express HY19730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Olmutinib (HM61713; BI-1482694) is an orally active and irreversible third EGFR tyrosine kinase inhibitor that binds to a cysteine residue near the kinase domain. Olmutinib is used for NSCLC.

Olmutinib (HM61713

Specifications

Chemical Name or Material Olmutinib
CAS 1353550-13-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H26N6O2S
Quantity 10 mg
Synonym HM61713, BI 1482694
Solubility Information DMSO : ≥ 44 mg/mL (90.43 mM)
SMILES C=CC(NC1=CC=CC(OC2=C3C(C=CS3)=NC(NC4=CC=C(N5CCN(C)CC5)C=C4)=N2)=C1)=O
Molecular Weight (g/mol) 486.59
Formula Weight 486.59
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.