Learn More
Olopatadine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Olopatadine Hydrochloride |
| CAS | 140462-76-6 |
| Molecular Formula | C21 H23 N O3 . Cl H |
| Quantity | 25 mg |
| Synonym | (11Z)-11-[3-(Dimethylamino)propylidene]-6,11-dihydrodibenz[b,e]oxepin-2-acetic Acid Hydrochloride (1:1), Dibenz[b,e]oxepin-2-acetic acid','11-[3-(dimethylamino)propylidene]-6,11-dihydro-','hydrochloride','(11Z)- (9CI), Dibenz[b,e]oxepin-2-acetic acid','11-[3-(dimethylamino)propylidene]-6,11-dihydro-','hydrochloride','(Z)-, (Z)-11-(3-Dimethylaminopropylidene)-6,11-dihydrodibenzo[b,e]oxepin-2-acetic acid hydrochloride, ALO 4943A, Allelock, Allerock, KW 4679, Olopatadine Hydrochloride, Olopax, Olopine, Patadine, Patanase, Pazeo |
| InChI Formula | InChI=1S/C21H23NO3.ClH/c1-22(2)11-5-8-18-17-7-4-3-6-16(17)14-25-20-10-9-15(12-19(18)20)13-21(23)24;/h3-4,6-10,12H,5,11,13-14H2,1-2H3,(H,23,24);1H/b18-8-; |
| SMILES | Cl.CN(C)CC\C=C/1\c2ccccc2COc3ccc(CC(=O)O)cc13 |
| IUPAC Name | 2-[(11Z)-11-[3-(dimethylamino)propylidene]-6H-benzo[c][1]benzoxepin-2-yl]acetic acid;hydrochloride |
| Molecular Weight (g/mol) | 373.87 |
| Formula Weight | 373.1445 |