Learn More
OMA1 Mouse anti-Human, Unconjugated, Clone: 1H4D12, Proteintech
Product Code. 16830086 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16830086 150 μL 150µL
16840086 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16830086 Supplier Proteintech Supplier No. 674491IG150UL
Mouse Monoclonal Antibody
The OMA1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human OMA1. This antibody recognizes human, mouse antigen. The OMA1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Antigen OMA1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1H4D12
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 115209
Target Species Human, Mouse
Isotype IgG1