missing translation for 'onlineSavingsMsg'
Learn More

OMA1 Mouse anti-Human, Unconjugated, Clone: 1H4D12, Proteintech

Product Code. p-7223785 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16840086 20 μL 20µL
16830086 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16840086 Supplier Proteintech Supplier No. 674491IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The OMA1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human OMA1. This antibody recognizes human, mouse antigen. The OMA1 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen OMA1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1H4D12
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 115209
Target Species Human, Mouse
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.