Learn More
Invitrogen™ Omalizumab Humanized Recombinant Human Monoclonal Antibody
GREENER_CHOICE
Human Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA541716
Product Code. 17788333
  • € 528.00 / 100µg
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Omalizumab with trade name Xolair, is an FDA-approved drug for the treatment of moderate to severe persistent asthma in adults and adolescents (12 years of age and above). It is a recombinant humanized monoclonal antibody. It selectively binds to human immunoglobulin E (IgE). Omalizumab inhibits the binding of IgE to the high-affinity IgE receptor (FcERI) on the surface of mast cells and basophils.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Omalizumab Humanized
Recombinant Monoclonal
Unconjugated
IGE25; olizumab; pSVIE26; rhuMab-E25; Xolair
Human IGHE.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
ELISA, SDS-Page
3.42 mg/mL
PBS with no preservative; pH 7.4
Human
Protein A/G
RUO
Human
Antibody
IgG1 κ