missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ombitasvir, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | Ombitasvir |
| CAS | 1258226-87-7 |
| Molecular Formula | C50H67N7O8 |
| Quantity | 2.5 mg |
| Synonym | ABT 267, 2,2'-[[(2S,5S)-1-[4-(1,1-Dimethylethyl)phenyl]-2,5-pyrrolidinediyl]di-4,1-phenylene]bis[N-(methoxycarbonyl)-L-valyl-L-prolinamide] |
| InChI Formula | InChI=1S/C50H67N7O8/c1-30(2)42(53-48(62)64-8)46(60)55-28-10-12-40(55)44(58)51-35-20-14-32(15-21-35)38-26-27-39(57(38)37-24-18-34(19-25-37)50(5,6)7)33-16-22-36(23-17-33)52-45(59)41-13-11-29-56(41)47(61)43(31(3)4)54-49(63)65-9/h14-25,30-31,38-43H,10-13,26-29H2,1-9H3,(H,51,58)(H,52,59)(H,53,62)(H,54,63)/t38-,39-,40-,41-,42-,43-/m0/s1 |
| SMILES | COC(=O)N[C@@H](C(C)C)C(=O)N1CCC[C@H]1C(=O)Nc2ccc(cc2)[C@@H]3CC[C@H](N3c4ccc(cc4)C(C)(C)C)c5ccc(NC(=O)[C@@H]6CCCN6C(=O)[C@@H](NC(=O)OC)C(C)C)cc5 |
| IUPAC Name | methyl N-[(2S)-1-[(2S)-2-[[4-[(2S,5S)-1-(4-tert-butylphenyl)-5-[4-[[(2S)-1-[(2S)-2-(methoxycarbonylamino)-3-methylbutanoyl]pyrrolidine-2-carbonyl]amino]phenyl]pyrrolidin-2-yl]phenyl]carbamoyl]pyrrolidin-1-yl]-3-methyl-1-oxobutan-2-yl]carbamate |
| Molecular Weight (g/mol) | 894.11 |
| Formula Weight | 893.5051 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction