missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OMD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111654
This item is not returnable.
View return policy
Description
Immunogen sequence: PGLPSSLMYL SLENNSISSI PEKYFDKLPK LHTLRMSHNK LQDIPYNIFN LPNIVELSVG HNKLKQAFYI P Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 86%.
OMD may be implicated in biomineralization processes. This protein has a function in binding of osteoblasts via the alpha(V)beta(3)-integrin.
Specifications
| OMD | |
| Polyclonal | |
| Unconjugated | |
| OMD | |
| Keratan sulfate proteoglycan osteomodulin; KSPG osteomodulin; Omd; OSAD; Osteoadherin; osteoadherin proteoglycan; osteomodulin; SLRR2C; UNQ190/PRO216; UNQ190/PRO216SLRR2C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4958 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99983 | |
| OMD | |
| Recombinant Protein corresonding to Human OMD. Recombinant protein control fragment (Product #RP-108560). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction