Learn More
Cytiva Omega Membrane Discs
Click to view available options
MWCO:
1 kDa
10 kDa
100 kDa
30 kDa
5 kDa
Diameter (Metric) Membrane:
43 mm
62 mm
76 mm
Unit Size:
Pack of 12
Description
- Polyethersulfone membrane provides higher flow rates and lower protein binding than competitive regenerated cellulose
- Lower processing times and the highest possible recoveries
- Discs can be washed and reused
- Replacement membranes for other stirred cell systems concentration, fractionation, purification, buffer exchange, or desalting of protein, cell broths, and other biomolecules
- For research use only
Specifications
Specifications
| Material (Membrane) | PES |
| MWCO | 30 kDa |
| Sterility | Non-sterile |
| No. per Pack | 12/Pk. |
| Diameter (Metric) Membrane | 43 mm |