Learn More
Diba Omnifit™ BenchMark™ Replacement O-rings for Column Assemblies
Product Code. 15835574
Encompass_Preferred
Click to view available options
Diameter (Metric) Outer:
15 mm
35 mm
50 mm
Unit Size:
Pack of 6
Product Code. 15835574
Brand: Diba 006OR35V
This item requires a price update prior to being ordered.
Request a Price for this item here.
Choose from FKM or FFKM O-rings
Use for quick-disconnect fittings, valves, pumps, and other fluid handling products.
Specifications
Type O-ring
Color Black
Diameter (English) Outer 1.37 in.
Diameter (Metric) Outer 35 mm
For Use With (Application) Chromatography
For Use With Omnifit Benchmark Chromatography Columns
Overall Length (Metric) 102 mm
Material FKM