Learn More
Diba Omnifit™ BenchMark™ Replacement O-rings for Column Assemblies
Product Code. 15895564
Encompass_Preferred
Click to view available options
Diameter (Metric) Outer:
15 mm
35 mm
50 mm
Unit Size:
Pack of 6
Product Code. 15895564
Brand: Diba 006OR15V
This item requires a price update prior to being ordered.
Request a Price for this item here.
Choose from FKM or FFKM O-rings
Use for quick-disconnect fittings, valves, pumps, and other fluid handling products.
Specifications
Type O-ring
Color Black
Diameter (Metric) Outer 15 mm
Diameter (Metric) Inner 15 mm
For Use With (Application) Chromatography
For Use With Omnifit Benchmark Chromatography Columns
Overall Length (Metric) 76 mm
Material FKM