Learn More
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with One Fixed and One Adjustable Endpieces
Product Code. 15895504
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Column Capacity:
0.1 to 0.7 mL
0.1 to 2.4 mL
0.2 to 5.5 mL
0.4 to 12.4 mL
1.2 to 34.4 mL
1.4 to 4.1 mL
108.0 to 147.3 mL
11.0 to 17.3 mL
134.7 to 211.7 mL
142.4 to 181.6 mL
17.3 to 23.6 mL
19.6 to 58.9 mL
191.4 to 230.7 mL
211.7 to 288.6 mL
22.8 to 29.1 mL
24.7 to 38.9 mL
279.0 to 356.0 mL
280.4 to 440.7 mL
3.1 to 9.4 mL
30.6 to 36.9 mL
38.5 to 115.5 mL
38.9 to 53.0 mL
4.8 to 7.5 mL
440.7 to 600.9 mL
51.2 to 65.4 mL
580.9 to 741.1 mL
68.7 to 108.0 mL
68.9 to 83.1 mL
7.1 to 21.2 mL
7.5 to 10.3 mL
781.2 to 941.4 mL
9.9 to 12.7 mL
Unit Size:
Each
32 product options available for selection
Product selection table with 32 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Column Capacity unitSize
15895504 400 mm 50 mm 580.9 to 741.1 mL Each
15805444 150 mm 10 mm 3.1 to 9.4 mL Each
15805474 500 mm 15 mm 68.9 to 83.1 mL Each
15805504 400 mm 35 mm 279.0 to 356.0 mL Each
15815434 330 mm 6.6 mm 7.5 to 10.3 mL Each
15815464 250 mm 15 mm 24.7 to 38.9 mL Each
15815494 150 mm 35 mm 38.5 to 115.5 mL Each
15825424 100 mm 6.6 mm 0.1 to 2.4 mL Each
15825454 500 mm 10 mm 30.6 to 36.9 mL Each
15825484 330 mm 25 mm 108.0 to 147.3 mL Each
15825514 500 mm 50 mm 781.2 to 941.4 mL Each
15835444 250 mm 10 mm 11.0 to 17.3 mL Each
15835474 100 mm 25 mm 1.2 to 34.4 mL Each
15835504 250 mm 50 mm 280.4 to 440.7 mL Each
15845434 400 mm 6.6 mm 9.9 to 12.7 mL Each
15845464 330 mm 15 mm 38.9 to 53.0 mL Each
15845494 250 mm 35 mm 134.7 to 211.7 mL Each
15865444 330 mm 10 mm 17.3 to 23.6 mL Each
15865474 150 mm 25 mm 19.6 to 58.9 mL Each
15865504 330 mm 50 mm 440.7 to 600.9 mL Each
15875434 100 mm 10 mm 0.2 to 5.5 mL Each
15875464 400 mm 15 mm 51.2 to 65.4 mL Each
15875494 330 mm 35 mm 211.7 to 288.6 mL Each
15885424 250 mm 6.6 mm 4.8 to 7.5 mL Each
15885454 150 mm 15 mm 7.1 to 21.2 mL Each
15885484 500 mm 25 mm 191.4 to 230.7 mL Each
15855424 150 mm 6.6 mm 1.4 to 4.1 mL Each
15855454 100 mm 15 mm 0.4 to 12.4 mL Each
15855484 400 mm 25 mm 142.4 to 181.6 mL Each
15895414 50 mm 6.6 mm 0.1 to 0.7 mL Each
15895444 400 mm 10 mm 22.8 to 29.1 mL Each
15895474 250 mm 25 mm 68.7 to 108.0 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
32 options
Product Code. 15895504 Supplier Diba Supplier No. 006EZS5040AF
Omnifit™ EZ SolventPlus™ Chromatography Columns with One Fixed and One Adjustable Endpieces feature PTFE, borosilicate glass, and Chemraz™ wetted materials for exceptional chemical resistance
  • PEEK™ column threads offer outstanding durability
  • Ergonomically designed adjustable endpiece offers quick and easy height adjustment
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Specifications
Temperature (Metric) Range 4 to 40°C
Diameter (Metric) Inner 50 mm
Type Liquid Chromatography Column
Max. Pressure 75 psi
Pore Size 10 and 30 μm
Column Capacity 580.9 to 741.1 mL
Length (Metric) 400 mm
Material Borosilicate Glass, PTFE, Chemraz™
Max. Temperature 40°C
Includes Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits) and a Click-N-Seal™ 10-32 PEEK conical port fitting.