Learn More
Diba Omnifit™ Frits for EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns
Product Code. 15885544
Encompass_Preferred
Click to view available options
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Material:
PTFE
Polyethylene
Pore Size:
5 μm
10 μm
20 μm
25 μm
30 μm
50 μm
Unit Size:
Pack of 20
Product Code. 15885544
Brand: Diba 006FR5030
This item requires a price update prior to being ordered.
Request a Price for this item here.
Omnifit™ Frits for EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns feature a range of porosities for your media and chemistry
Specifications
For Use With (Equipment) Omnifit™ EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns
Diameter (Metric) Inner 50 mm
Type Liquid Chromatography Column
Pore Size 30 μm
Length (Metric) 50 mm
Material PTFE