Learn More
Diba Omnifit™ Frits for EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns
Click to view available options
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Material:
PTFE
Polyethylene
Pore Size:
5 μm
10 μm
20 μm
25 μm
30 μm
50 μm
Unit Size:
Pack of 20
Specifications
Specifications
| For Use With (Equipment) | Omnifit™ EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns |
| Diameter (Metric) Inner | 6.6 mm |
| Type | Liquid Chromatography Column |
| Pore Size | 50 μm |
| Length (Metric) | 6 mm |
| Material | PTFE |