Learn More
Diba Omnifit™ Packing Sleeves for EZ and EZ SolventPlus™ Columns
Product Code. 15825274
Encompass_Preferred
Click to view available options
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Unit Size:
Each
Product Code. 15825274
Brand: Diba 006EPS06
This item requires a price update prior to being ordered.
Request a Price for this item here.
Use Omnifit™ Packing Sleeves for EZ and EZ SolventPlus™ Columns to join two columns for filling and packing
Specifications
For Use With (Equipment) Omnifit™ EZ and EZ SolventPlus™ Columns
Diameter (Metric) Inner 6.6 mm
Type Liquid Chromatography Column
Length (Metric) 4 cm