Learn More
Diba Omnifit™ Replacement Endpieces for EZ SolventPlus™ Columns
Product Code. 15805314
Encompass_Preferred
Click to view available options
Endcapped:
Adjustable
Fixed
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Unit Size:
Each
Product Code. 15805314
Brand: Diba 006ESAE06
This item requires a price update prior to being ordered.
Request a Price for this item here.
Select from fixed or adjustable Omnifit™ Replacement Endpieces for EZ SolventPlus™ Columns
Specifications
Endcapped Adjustable
For Use With (Equipment) Omnifit™ EZ SolventPlus™ Columns
Diameter (Metric) Inner 6.6 mm
Type Liquid Chromatography Column
Length (Metric) 15 cm
Material PTFE