Learn More
B Braun™ Omnifix™ Solo 3-Piece Syringes
Product Code. 10278554
Click to view available options
Volume (Metric):
3 mL
5 mL
20 mL
50 mL
Unit Size:
Each
Pack of 100
Product Code. 10278554
Brand: B Braun™ 4617022V
Edge
Features smooth plunger action and double-seal stopper. B Braun™ Omnifix™ Solo 3-Piece Syringes are designed for consistent performance.
  • High visibility; transparent barrel with permanent bold graduations
  • Secure backstop to prevent accidental plunger withdrawal
  • Double seal stopper prevents medication leakage
  • Large, stable finger grips
  • Components do not contain DEHP, PVC, or other rubber tree latex
  • Disposable
  • Piston with double sealing ring provides slow aspiration and injection
Specifications
Certifications/Compliance EN ISO 7886-1, ISO 10993 resp. DIN EN 30993
Material Polyisoprene Rubber (Tip), Polypropylene (Barrel), Polypropylene (Plunger)
Color Natural
Description High contrast black graduation
For Use With (Application) For slow aspiration and injection
Latex Free Yes
Product Type Centric Luer-lock 3-part Syringe
Disposable Single-use
Graduations 0.1 mL
Quantity 100/Pk.
Volume (Metric) 3 mL
Connection Type Centric Luer Lock
Graduated Yes
Sterility Sterile