missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ omp38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144424
This item is not returnable.
View return policy
Description
omp38 Polyclonal Antibody for Western Blot, ELISA
Porin. Induces apoptosis in human cells through caspases-dependent and AIF-dependent pathways. Purified Omp38 enters the cells and localizes to the mitochondria, which leads to a release of proapoptotic molecules such as cytochrome c and AIF (apoptosis-inducing factor).
Specifications
| omp38 | |
| Polyclonal | |
| Unconjugated | |
| omp38; Outer membrane protein Omp38 | |
| Recombinant Acinetobacter baumannii Outer membrane protein Omp38 (omp38) (20-356aa). | |
| 100 μL | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.61 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction