missing translation for 'onlineSavingsMsg'
Learn More

ON123300, MedChemExpress

Product Code. 16471477
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16471477

Brand: Medchem Express HY12624

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ON123300, a strong and brain-penetrant multi-kinase inhibitor, inhibits CDK4 (IC50=3.9 nM), Ark5 (IC50=5 nM), PDGFRβ (IC50=26 nM), FGFR1 (IC50=26 nM), RET (IC50=9.2 nM), and FYN (IC50=11 nM). Single agent ON123300 causes a dose-dependent suppression of phosphorylation of Akt as well as activation of Erk in brain tumors. ON123300 inhibits CDK6 with an IC50 of 9.82 nM.

ON123300, a strong and brain-penetrant multi-kinase inhibitor, inhibits CDK4 (IC50=3.9 nM), Ark5 (IC50=5 nM), PDGFRβ (IC50=26 nM), FGFR1 (IC50=26 nM), RET (IC50=9.2 nM), and FYN (IC50=11 nM). Single agent ON123300 causes a dose-dependent suppression of phosphorylation of Akt as well as activation of Erk in brain tumors. ON123300 inhibits CDK6 with an IC50 of 9.82 nM.

Specifications

Chemical Name or Material ON123300
Physical Form Solid
CAS 1357470-29-1
Color Orange
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C24H27N7O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 16.67 mg/mL (38.81 mM; ultrasonic and warming and heat to 60°C)
SMILES N#CC1=CC2=CN=C(NC3=CC=C(N4CCN(C)CC4)C=C3)N=C2N(C5CCCC5)C1=O
Molecular Weight (g/mol) 429.52
Formula Weight 429.52
Percent Purity 99.34%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.