Learn More
Invitrogen™ Onartuzumab Humanized Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA541934
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Onartuzumab has been used in trials studying the treatment of Neoplasms, Lung Cancer, Glioblastoma, Gastric Cancer, and Colorectal Cancer, among others.
Specifications
| Onartuzumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MetMAb; OA-5D5; PRO 143966 | |
| Human MET/RCCP2. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 2 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 F(ab) κ |