missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oncostatin M/OSM Antibody (17022), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2951J
This item is not returnable.
View return policy
Description
Oncostatin M/OSM Monoclonal antibody specifically detects Oncostatin M/OSM in Human samples. It is validated for Neutralization, Flow Cytometry, CyTOF
Specifications
| Oncostatin M/OSM | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Neutralization, Flow Cytometry, CyTOF | |
| 17022 | |
| Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| MGC20461, oncostatin M, oncostatin-M | |
| E. coli-derived recombinant human Oncostatin M/OSM, Ala26-Arg221, Accession # P13725 | |
| 0.5 mL | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 5008 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction