missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oncostatin M/OSM Antibody (OTI2B6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73146
This item is not returnable.
View return policy
Description
Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Oncostatin M/OSM | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:1000, Immunohistochemistry 1:50 | |
| MGC20461, oncostatin M, oncostatin-M | |
| Mouse | |
| 25.7 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2B6 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| OSM | |
| Recombinant protein expressed in E.coli corresponding to amino acids 25-252 of human OSM | |
| Affinity Purified | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 5008 | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction