missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oncostatin M/OSM Antibody (OTI2B6), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73146PCP
This item is not returnable.
View return policy
Description
Oncostatin M/OSM Monoclonal antibody specifically detects Oncostatin M/OSM in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Oncostatin M/OSM | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI2B6 | |
| Western Blot, Immunohistochemistry | |
| MGC20461, oncostatin M, oncostatin-M | |
| Recombinant protein expressed in E.coli corresponding to amino acids 25-252 of human OSM | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 5008 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction