Learn More
Lenz Laborglasintrumente One-Way Reaction/Distillation Units
Description
- Batch reactor for stirring under normal pressure or vacuum, with possibility of addition of liquids and boiling under reflux.
- The descending condenser with intermediate and final receiver (suitable for vacuum operation) allows the separation of the solvent from the product by one-way distillation after reaction.
- High-quality system components: Reaction vessels (6, 10, 20 or 30 l) with thermostatic jacket (heatable by a thermostatic bath), with bottom discharge valve (PTFE, no dead volume), made of DURAN™ tubing
- Flat flange lid (LF 150 or LF 200) 1 center neck NS 45/40, 2 side necks 45° NS 29/32, 1 side neck straight NS 14/23, material: DURAN™
- Feeding funnel Capacity: 1000 or 4000 ml, with graduation, NS 29/32, made of DURAN™ tubing Spiral condenser NS 29/32, S 29, jacket length: 300 mm, made of DURAN™ tubing Stirrer guide and stirrer with 16 mm shaft, stirrer: steel, PTFE coated
Specifications
Specifications
| Includes | Lab Flange 150 LF |
| Material | Duran Glass |
| Capacity (Metric) | 6 L |