Learn More
Lenz Laborglasintrumente One-Way Reaction/Distillation Units
Product Code. 11953591
Change view
Click to view available options
Capacity (Metric):
6 L
10 L
20 L
Includes:
Lab Flange 150 LF
Lab Flange 200 LF
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Includes unitSize
11953591 20 L Lab Flange 150 LF Each
11923591 6 L Lab Flange 150 LF Each
11933591 10 L Lab Flange 150 LF Each
11943591 6 L Lab Flange 200 LF Each
11963591 10 L Lab Flange 200 LF Each
11973591 20 L Lab Flange 200 LF Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11953591 Supplier Lenz Laborglasintrumente Supplier No. 06265494
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Batch reactor for stirring under normal pressure or vacuum, with possibility of addition of liquids and boiling under reflux.
  • The descending condenser with intermediate and final receiver (suitable for vacuum operation) allows the separation of the solvent from the product by one-way distillation after reaction.
  • High-quality system components: Reaction vessels (6, 10, 20 or 30 l) with thermostatic jacket (heatable by a thermostatic bath), with bottom discharge valve (PTFE, no dead volume), made of DURAN™ tubing
  • Flat flange lid (LF 150 or LF 200) 1 center neck NS 45/40, 2 side necks 45° NS 29/32, 1 side neck straight NS 14/23, material: DURAN™
  • Feeding funnel Capacity: 1000 or 4000 ml, with graduation, NS 29/32, made of DURAN™ tubing Spiral condenser NS 29/32, S 29, jacket length: 300 mm, made of DURAN™ tubing Stirrer guide and stirrer with 16 mm shaft, stirrer: steel, PTFE coated
Specifications
Includes Lab Flange 150 LF
Material Duran Glass
Capacity (Metric) 20 L