Learn More
Invitrogen™ ONECUT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114396
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human HEK293 whole cell, human HepG2 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
ONECUT2 is a member of the transcription factors of the ONECUT class, whose prototype is hepatocyte nuclear factor (HNF)-6. The distribution of OC-2 mRNA in humans is tissue-restricted, the strongest expression being detected in the liver and skin. ONECUT2 participates in the network of transcription factors required for liver differentiation and metabolism.
Specifications
| ONECUT2 | |
| Polyclonal | |
| Unconjugated | |
| ONECUT2 | |
| C730009D12; Hepatocyte nuclear factor 6-beta; HNF6B; HNF-6B; HNF-6-beta; OC2; OC-2; One cut domain family member 2; one cut domain, family member 2; one cut homeobox 2; onecut 2; ONECUT2; ONECUT-2 homeodomain transcription factor; RGD1564677; si:ch1073-41k10.1; transcription factor ONECUT2; transcription factor ONECUT-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 225631, 9480 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| O95948, Q6XBJ3 | |
| ONECUT2 | |
| A synthetic peptide corresponding to a sequence of human ONECUT2 (MKAAYTAYRCLTKDLEGCAMNPELTMESLGTLH) (aa 1-33). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.