missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ONECUT3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15807303
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15807303 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15807303 Supplier Invitrogen™ Supplier No. PA568887

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 90%; Rat: 100%; Zebrafish: 79%.

Transcriptional activator. Binds the consensus DNA sequence 5'-DHWATTGAYTWWD-3' on a variety of gene promoters such as those of HNF3B and TTR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ONECUT3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ONECUT3
Gene Accession No. Q8K557
Gene Alias OC3; OC-3; One cut domain family member 3; one cut domain, family member 3; one cut homeobox 3; Onecut3; Transcription factor ONECUT-3
Gene Symbols ONECUT3
Host Species Rabbit
Immunogen synthetic peptide directed towards the c terminal region of mouse ONECUT3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246086
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.