Learn More
Kolmi Hopen™ Op-Air™ One Anti-glare Medical Mask with Foam Band
Product Code. 17740189 Shop All Medicom Products
Change view
Click to view available options
:
Op-Air One Anti-glare Medical Mask with Foam Band; Certifications/Compliance (Europe): (EU) 2017/745, EN 14683, ISO 13485, ISO 9001; Color: Blue; Disposable: Yes; Fastening Type: Ties; Material: Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17740189 Op-Air One Anti-glare Medical Mask with Foam Band; Certifications/Compliance (Europe): (EU) 2017/745, EN 14683, ISO 13485, ISO 9001; Color: Blue; Disposable: Yes; Fastening Type: Ties; Material: Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17740189 Supplier Kolmi Hopen™ Supplier No. M24101MN30
Edge
Be prepared for long hours of wear with this anti-glare, anti-fog medical mask complete with a comfortable foam band and anti-reflective tape.
  • High filtration media (polypropylene)
  • Layers (interior to exterior): wet laid cellulose, melt blown polypropylene, spunbonded polypropylene
  • Soft, non-woven mask ties and edges
  • Anatomical folding limits exposure to face
  • Integrated nose bar and foam strip to reduce glasses fogging
  • Black, anti-reflective tape limits glare phenomena in operating room
  • Fiberglass-free filter
  • Produced in ISO 8 clean room (particle and microbiological control)
Specifications
Color Blue
Fastening Type Ties
Material Polypropylene
Certifications/Compliance (Europe) (EU) 2017/745, EN 14683, ISO 13485, ISO 9001
Disposable Single-use
For Use With (Application) Medical
Packaging Quantity 50 Pk.