Learn More
Kolmi Hopen™ Op-Air™ PRO Oxygen Type IIR FFP2 Respiratory Mask
This respiratory pouch mask is ideal for protection against infectious diseases and in dusty environments.
Brand: Kolmi Hopen™ M52214S-WH
17.01 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Additional Details : Weight : 4.75000kg
Description
- Filtering half mask with FFP2 NR D efficiency class
- Developed to protect against solid and liquid aerosols and particles
- Protects environment against droplets emitted by wearer and protects wearer from potential body fluid splashes
- Knitted head loops are ultrasonically welded for optimal comfort
- Fits perfectly to face to prevent exposure
- Made in France
Specifications
(EU) 2017/745, EN 14683:2019+AC:2019 | |
Pouch/Duckbill | |
Polypropylene | |
Yes | |
50 Pk. |
White | |
Single-use | |
Small | |
Polybags |