missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPA3 Antibody (OTI3G4), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73147AF700
This item is not returnable.
View return policy
Description
OPA3 Monoclonal specifically detects OPA3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| OPA3 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| OPA3 | |
| Full length recombinant protein of human OPA3 (NP_079412) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3G4 | |
| Western Blot, Immunohistochemistry | |
| FLJ22187, FLJ25932, MGA3Optic atrophy 3 (Iraqi-Jewish ′optic atrophy plus′), MGC75494, optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia), optic atrophy 3 protein | |
| Mouse | |
| Affinity Purified | |
| Vision | |
| 80207.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction