missing translation for 'onlineSavingsMsg'
Learn More

OPA3 Monoclonal Antibody (OTI1G3), TrueMAB™, OriGene

Product Code. 17571724
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17571724 100 μL 100µL
17591724 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17571724 Supplier Origene Technologies Supplier No. TA809469

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The mouse ortholog of this protein co-purifies with the mitochondrial inner membrane. Mutations in this gene have been shown to result in 3-methylglutaconic aciduria type III and autosomal domit optic atrophy and cataract. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen OPA3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene OPA3
Gene Accession No. Q9H6K4
Gene Alias MGA3
Gene Symbols OPA3
Host Species Mouse
Immunogen Full length human recombit protein of human OPA3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80207
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.