missing translation for 'onlineSavingsMsg'
Learn More

OPA3 Monoclonal Antibody (OTI1G3), TrueMAB™, OriGene

Product Code. p-200013663
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17591724 30 μL 30µL
17571724 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17591724 Supplier Origene Technologies Supplier No. TA809469S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

OPA3 Monoclonal antibody specifically detects OPA3 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen OPA3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CTSL
Gene Accession No. P07711
Gene Alias D630048P19Rik; FLJ22187; FLJ25932; Gm1425; Gm472; MGA3; MGC75494; OPA3; OPA3 outer mitochondrial membrane lipid metabolism regulator; optic atrophy 3; optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia); Optic atrophy 3 (Iraqi-Jewish 'optic atrophy plus'); optic atrophy 3 protein; Optic atrophy 3 protein homolog; outer mitochondrial membrane lipid metabolism regulator OPA3
Host Species Mouse
Immunogen Full length human recombinant protein of human CTSL1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1514
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.