missing translation for 'onlineSavingsMsg'
Learn More

OPA3 Mouse anti-Human, Clone: OTI3G4, TrueMAB™, Origene

Product Code. 17501634
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
Unit Size:
30µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17501634 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17501634 Supplier Origene Technologies Supplier No. TA809426S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

OPA3 Monoclonal antibody specifically detects OPA3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen OPA3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene OPA3
Gene Accession No. Q9H6K4
Gene Alias D630048P19Rik; FLJ22187; FLJ25932; Gm1425; Gm472; MGA3; MGC75494; OPA3; OPA3 outer mitochondrial membrane lipid metabolism regulator; optic atrophy 3; optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia); Optic atrophy 3 (Iraqi-Jewish 'optic atrophy plus'); optic atrophy 3 protein; Optic atrophy 3 protein homolog; outer mitochondrial membrane lipid metabolism regulator OPA3
Host Species Mouse
Immunogen Full length human recombinant protein of human OPA3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80207
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.