missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OPCML Polyclonal Antibody
GREENER_CHOICE

Product Code. 17792224
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17792224 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17792224 Supplier Invitrogen™ Supplier No. PA5120831

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, 293T, HeLa, Mouse brain, Mouse pancreas, Mouse kidney, Rat brain, Rat pancreas, Rat kidney. The target is usually found in the following locations: Cell membrane, GPI-anchor, Lipid-anchor. Immunogen sequence: ASAVPMAEFQ WFKEETRLAT GLDGMRIENK GRMSTLTFFN VSEKDYGNYT CVATNKLGNT NASITLYGPG AVIDGVNSAS RALACLWLSG TLLAHFFIKF.

Binds opioids in the presence of acidic lipids; probably involved in cell contact.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OPCML
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene OPCML
Gene Accession No. P32736, Q14982
Gene Alias 2900075O15Rik; 3732419F12; AI844366; B930023M13Rik; C230027C17; Gm181; IgLON family member 1; IGLON1; OBCAM; OPCM; OPCML; opioid binding cell adhesion molecule; opioid binding protein/cell adhesion molecule like; opioid binding protein/cell adhesion molecule-like; opioid binding protein/cell adhesion molecule-like preprotein; opioid-binding cell adhesion molecule; opioid-binding protein/cell adhesion molecule; opioid-binding protein/cell adhesion molecule-like
Gene Symbols OPCML
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 246-345 of human OPCML (NP_002536.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116597, 330908, 4978
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.